missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D(-)-Aspartic acid, +99%
Chemical Identifiers
CAS | 1783-96-6 |
---|---|
Molecular Formula | C4H7NO4 |
Molecular Weight (g/mol) | 133.103 |
MDL Number | MFCD00063081 |
InChI Key | CKLJMWTZIZZHCS-UWTATZPHSA-N |
Synonym | d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d |
PubChem CID | 83887 |
ChEBI | CHEBI:17364 |
IUPAC Name | (2R)-2-aminobutanedioic acid |
SMILES | C(C(C(=O)O)N)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10574274
|
Thermo Scientific Chemicals
204260050 |
5 g | Glass Bottle |
N/A
|
N/A | ||||
10296540
|
Thermo Scientific Chemicals
204260250 |
25 g | Glass Bottle |
N/A
|
N/A | ||||
10145403
|
Thermo Scientific Chemicals
204261000 |
100 g | Glass Bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1783-96-6 | |
133.103 | |
CKLJMWTZIZZHCS-UWTATZPHSA-N | |
83887 | |
(2R)-2-aminobutanedioic acid |
C4H7NO4 | |
MFCD00063081 | |
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d | |
CHEBI:17364 | |
C(C(C(=O)O)N)C(=O)O |
Specifications
>300.0°C | |
White | |
C4H7NO4 | |
HO2CCH2CH(NH2)CO2H | |
04, 471 | |
15, 830 | |
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d | |
CKLJMWTZIZZHCS-UWTATZPHSA-N | |
(2R)-2-aminobutanedioic acid | |
− 25.80 | |
83887 | |
133.1 | |
− 25.80 (20.00°C c=5,5N HCl) | |
D(-)-Aspartic acid, 99+% |
1783-96-6 | |
99+% | |
Authentic | |
MFCD00063081 | |
0.4% max. (105°C, 3 hrs) | |
0.3% max. (TLC) | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol and ether | |
C(C(C(=O)O)N)C(=O)O | |
0.1% max. | |
133.103 | |
CHEBI:17364 | |
≥99% | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 217-234-6
missing translation for 'rtecsNumber' : CI9097500
RUO – Research Use Only