missing translation for 'onlineSavingsMsg'
Learn More

Cytochalasin D, Tocris Bioscience™

Product Code. 18782574
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Disrupts actin filament function

Potent disruptor of actin filament function. Alters tight junction permeability. Unlike cytochalasin B (Cat. No. 5474), does not inhibit monosaccharide transport across the plasma membrane.

Chemical Identifiers

CAS 22144-77-0
Molecular Formula C30H37NO6
Molecular Weight (g/mol) 507.627
InChI Key SDZRWUKZFQQKKV-DKLHZCHASA-N
Synonym cytochalasin d
PubChem CID 131668624
SMILES CC1CC=CC2C(C(=C)C(C3C2(C(C=CC(C1=O)(C)O)OC(=O)C)C(=O)NC3CC4=CC=CC=C4)C)O

Specifications

Quantity 1 mg
Formula Weight 507.63
Percent Purity >95%
Chemical Name or Material Cytochalasin D
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.