Learn More
R&D Systems™ Cynomolgus Monkey IL-3R alpha (EHH61867.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC3159
Product Code. 18783277
  • 318.00€ / 10µg
Explore more special offers
Specifications
IL-3 R alpha/CD123
10 μg at 0.2 μg/μL
1150 bp
CD123, CD123 antigen, hIL3Ra, hIL-3 Ra, IL-3 R alpha, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3 R subunit alpha, IL3RA, IL-3 RA, IL-3 R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Cynomolgus Monkey
EHH61867.1
cDNA is provided in 10 mM Tris-Cl, pH 8.5
3563
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.