Learn More
R&D Systems™ Cynomolgous Monkey Hepassocin (EHH64014.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand: R&D Systems™ RDC3087
This item is not returnable.
View return policy
Specifications
Hepassocin/FGL1 | |
10 μg at 0.2 μg/μL | |
952 bp | |
FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1 | |
Cynomolgus Monkey |
EHH64014.1 | |
cDNA is provided in 10 mM Tris-Cl, pH 8.5 | |
2267 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |