Learn More
R&D Systems™ Cynomolgous Monkey Hepassocin (EHH64014.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC3087
Product Code. 18773777
  • 322.00€ / 10µg
Explore more special offers
Specifications
Hepassocin/FGL1
10 μg at 0.2 μg/μL
952 bp
FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1
Cynomolgus Monkey
EHH64014.1
cDNA is provided in 10 mM Tris-Cl, pH 8.5
2267
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.