missing translation for 'onlineSavingsMsg'
Learn More

CYM 50769, Tocris Bioscience™

Product Code. 30001985
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Novel non-peptide NPBWR1 antagonist

Novel non-peptide antagonist of neuropeptide W/B receptor 1 (NPBWR1, GPR7) (IC50 = 0.12 μM).

Chemical Identifiers

CAS 1421365-63-0
Molecular Formula C24H17ClN2O3
Molecular Weight (g/mol) 416.861
InChI Key QHVSQUYCVUHYKT-UHFFFAOYSA-N
Synonym cym hplc, 5-chloro-2-9h-fluoren-9-yl-4-4-methoxyphenoxy pyridazin-3-one, 5-chloro-2-9h-fluoren-9-yl-4-4-methoxyphenoxy-3 2h-pyridazinone
PubChem CID 50904505
IUPAC Name 5-chloro-2-(9H-fluoren-9-yl)-4-(4-methoxyphenoxy)pyridazin-3-one
SMILES COC1=CC=C(C=C1)OC2=C(C=NN(C2=O)C3C4=CC=CC=C4C5=CC=CC=C35)Cl

Specifications

Color White
Quantity 10 mg
Formula Weight 416.86
Percent Purity >98%
Physical Form Solid
Chemical Name or Material CYM 50769
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'