Learn More
Kartell Cylindrical Test Tube with Screw Cap, Polypropylene
Test tubes with quick closure screw caps (PE). Leak-proof, and suitable for centrifugation. The design of the cap avoids any contamination due to drops of liquid on the inner surface.
Brand:  Kartell 8830306
 View more versions of this product
Product Code. 18078031
  • 194.00€ / Pack of 1000
Explore more special offers
Description
Description
  • For laboratory use
  • For collection and evaluation of samples
  • Caps are not autoclavable
Specifications
Specifications
Yellow
15 mL
Polypropylene
Single-use
Autoclavable
Screw Cap
Clear
100 mm
Cylindrical
Non-sterile
No
Centrifuge Tube