accessibility menu, dialog, popup
Test
chevron-down
Select Language
English
French
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Hotplates and Stirrers
Stirring Bars and Rods
11517792
Print
Fisherbrand™ Cylindrical PTFE Stir Bars
Dimensions (Dia. x L): 3 dia. x 10mmL
For use with small containers
Brand:
Fisherbrand™ 001.810
Product Code.
11517792
7.40€
/ Pack of 10
Quantity
In Stock
Log in
to see stock.
Explore more special offers
Description
Made of PTFE
Specifications
Specifications
Dimensions (L x W x H)
10 x 3mm
Diameter (Metric)
3 mm
Length (Metric)
10 mm
Quantity
10/Pk.