Learn More
Draeger Safety Cyclohexane Detection Tube
Draeger Safety Cyclohexane Detection Tube features a dynamic measuring range and lower sensitivity.
Brand:  Draeger Safety 8103671
Product Code. 15604749
  • 143.00€ / Pack of 10
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Explore more special offers
Description
Description
  • Lower sensitivity starting at 40 ppm
  • Dynamic measurement range of 40-3,000 ppm, with two scales
  • n=5 with a scale of 40-200 ppm (approx. 75 seconds)
  • n=1 with a scale of 300-3,000 ppm (only 15 seconds)
  • Much shorter measuring time
  • Expanded humidity range, 1-35 mg/L
  • Extended operating temperature range, 0-40°C
  • More distinct color change, white to greenish brown
Specifications
Specifications
Detects cyclohexane 40/A
40 to 200 ppm, 300 to 3000 ppm
10/Box
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • drager tubes cyclohexane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.