Learn More
Draeger Safety Cyclohexane Detection Tube
Draeger Safety Cyclohexane Detection Tube features a dynamic measuring range and lower sensitivity.
Brand: Draeger Safety 8103671
This item is not returnable.
View return policy
Description
- Lower sensitivity starting at 40 ppm
- Dynamic measurement range of 40-3,000 ppm, with two scales
- n=5 with a scale of 40-200 ppm (approx. 75 seconds)
- n=1 with a scale of 300-3,000 ppm (only 15 seconds)
- Much shorter measuring time
- Expanded humidity range, 1-35 mg/L
- Extended operating temperature range, 0-40°C
- More distinct color change, white to greenish brown
Specifications
Detects cyclohexane 40/A | |
40 to 200 ppm, 300 to 3000 ppm | |
10/Box |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
- drager tubes cyclohexane
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.