missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane ≥ 99.5% (by GC), ChromAR™ for HPLC, Macron Fine Chemicals™
Click to view available options
2.5 L
Description
- Ultraviolet Absorbance (1.00-cm cell vs water)
- 210 nm ≤ 0.4
- 220 nm ≤ 0.1
- 230 nm ≤ 0.05
- 240 nm ≤ 0.01
- 350 nm ≤ 0.004
Specifications
Specifications
CAS | 110-82-7 |
Quantity | 2.5 L |
Flash Point | -18°C |
Assay Percent Range | ≥ 99.5% |
Residue after Evaporation | ≤ 0.0005% |
Molecular Formula | C6H12 |
Titratable Acid | ≤ 0.0005 meq/g |
MDL Number | MFCD00003814 |
UN Number | 1145 |
Water | ≤ 0.01% |
Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P331-Do NOT induce vomiting.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'