missing translation for 'onlineSavingsMsg'
Learn More

α-Cyclodextrin (White Powder), Fisher BioReagents

Product Code. p-25246
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10695215

Brand: Fisher BioReagents BP25015

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
  • A cyclic polysaccharide comprised of 6 glucose units that improves the solubility of hydrophobic molecules
  • Improves the solubility of nonpolar molecules such as fatty acids, lipids, and cholesterol in aqueous media
  • Also used for selective precipitation of enantiomeric, positional or structural isomers in chromatrographic separations and purification methods
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.846
InChI Key HFHDHCJBZVLPGP-UHFFFAOYSA-N
Synonym cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin
PubChem CID 320760
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O

Specifications

Melting Point 278°C
Color White
Loss on Drying 13% max.
Packaging Amber Glass
Quantity 5 g
Merck Index 15, 2710
Specific Rotation 143 to 153° (+ or -)
Formula Weight 972.9
Physical Form Powder or Solid
Chemical Name or Material α-Cyclodextrin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.