Learn More
WTW™ Cyanide Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand:  WTW™ 250492
 View more versions of this product
Product Code. 10009670
  • 178.00€ / Each
Explore more special offers
Specifications
DIN 38 405 D13/ISO 6703
Measurement of Cyanide in wastewater from electroplating and metal processing works, drinking water
Cuvettes measuring 10mm, 20mm and 50mm
0.002 to 0.500 mg/L
25 mL
PhotoLab S12, PhotoLab Spectral
100 Pack
Test powder