Learn More
3M™ Safety-Walk™ Cushion Matting 5270E
Product Code. 17128740
Click to view available options
Length (Metric):
3.1 m
6.1 m
9.1 m
Unit Size:
1 roll
Each
Product Code. 17128740
Brand: 3M™ Safety-Walk™ 7100116722
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This motor oil resistant mat, suitable for heavy foot traffic in dry areas, provides a comfortable surface for walking or standing.
  • Coiled cushion matting compresses evenly across the bottom of the shoe, reducing leg and back fatigue
  • Textured top improves traction for safer footing
  • Suitable for dry work areas such as assembly lines and production areas
  • Supplied with edging to allow for wheeled traffic
  • Withstands most industrial and cleaning chemicals
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Length (English) 3 ft.
Length (Metric) 9.1 m
Type Cushion Mat
Width (English) 2 ft.
Width (Metric) 0.61 m