Learn More
Greiner Bio-One Culture Tubes with Two-Position Vent Stoppers
Product Code. 10018820
Click to view available options
Capacity (Metric):
5 mL
10 mL
14 mL
Diameter (Metric):
12 mm
17 mm
18 mm
Quantity:
1000 Pack
2000 Pack
Unit Size:
Pack of 1000
Pack of 2000
Product Code. 10018820
Brand: Greiner Bio-One 184261
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Manufactured from polypropylene for excellent thermal, mechanical and chemical resistance. Greiner Bio-One™ Culture Tubes with Two-Position Vent Stoppers are ideal for storage of chemical and biological samples. two-position top allows ventilation and secure fit.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Two-position vent stopper fulfills two functions:
    • Attach lightly for uniform ventilation of the tube contents 
    • Attache firmly, by pressing in the stopper further, for an airtight closure of the tube
  • Graduations and writing area
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN ISO 10993
Color Natural
Capacity (Metric) 10 mL
Material Polypropylene
Disposable Single-use
Quantity 1000 Pack
Sterility Sterile
Graduated Yes
Diameter (Metric) 17 mm
Closure Type Vent Stopper