Learn More
Greiner Bio-One Culture Tube with Two-Position Vent Cap
Click to view available options
Packaging:
1/Bag; 750/Cs.
25; 1000
Unit Size:
Pack of 1000
Pack of 750
Description
Sterile
Specifications
Specifications
| Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens |
| Closure Color | Translucent |
| Closure Material | LDPE |
| Color | Clear |
| Material | Polystyrene |
| Packaging | 25; 1000 |
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
| Closure Type | Vent Stopper |