Learn More
Sartorius Cubis™ II Precision Lab Balance
BIOPHARMA
Product Code. p-7238110 Shop All Sartorius Products
Click to view available options
Capacity:
10,200 g
1010 g, 2200 g
1200 g
14,200 g
150 g, 300 g, 620 g
1500 g, 3000 g, 6200 g
2200 g
320 g
3500 g, 7000 g, 14,200 g
5200 g
8200 g
Readability:
1 mg
1 mg, 10 mg
1 mg, 2 mg, 5 mg
10 mg
10 mg, 20 mg, 50 mg
100 mg
Model:
Balance with Draft Shield A
Balance with Draft Shield E
Balance with Draft Shield I
Balance with Draft Shield R
Balance with Draft Shield U
Balance with Draft Shield U and without QP99 Software
Balance without Draft Shield
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 16303297
Brand: Sartorius MCA6202P2S000 QP99
Regulatory: This product is under eco-contribution tax.
Ideal model for every application with a maximum load between 320 g and 14,200 g and a readability of 1 mg to 100 mg.
The New Generation of Modular Premium Laboratory Balances
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series from Sartorius.
All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Balance with Type Approval Certificate for China
Connection Type D-sub connector, 9-pin
Capacity 1500 g, 3000 g, 6200 g
Display Type Color Touchscreen
Readability 10 mg, 20 mg, 50 mg
Includes Internal Calibration
Material Aluminum, Plastic PBT, Optiwhite Float Glass, and Stainless Steel
Model Balance without Draft Shield
Stabilization Time 1 sec.
Electrical Requirements 100/240 V (± 10%), 50/60 Hz, 1.0 A