Learn More
Sartorius Cubis™ II Precision Lab Balance
BIOPHARMA
Product Code. p-7237937 Shop All Sartorius Products
Click to view available options
Capacity:
1010 g, 2200 g
12,200 g
1200 g
14,200 g
2200 g
3200 g
4200 g, 32200 g
620 g
6200 g
8200 g
Readability:
1 mg
10 mg
10 mg, 100 mg
100 mg
Model:
Draft Shield E: Manual glass draft shield for precision balances
Draft Shield R: Flat, stainless steel weighing pan draft shield (removable, with no glass components) for all precision balances
Draft shield U: Manual glass
Weighing pan without draft shield
Unit Size:
1 unit
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 16213252
Brand: Sartorius MCE12201S0CEU
Regulatory: This product is under eco-contribution tax.
Maximum load between 320 g and 14,200 g and a readability of 1 mg to 100 mg provide, ideal model for every application.
The New Generation of Modular Premium Laboratory Balances
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series from Sartorius.
All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Balance with Type Approval Certificate for China
Connection Type D-sub connector, 9-pin
Capacity 12,200 g
Display Type Color Touchscreen
Readability 100 mg
Material Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
Model Weighing pan without draft shield
Stabilization Time 0.8 sec.
Electrical Requirements 100 to 240 VAC (± 10%), 50/60 Hz, 1.0 A