missing translation for 'onlineSavingsMsg'
Learn More

CU CPT 4a, Tocris Bioscience™

Product Code. 30002005
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective TLR3 inhibitor

Selective TLR3 inhibitor (IC50 = 3.44 μM in RAW 264.7 cells); suppresses downstream signaling pathways mediated by the TLR3/dsRNA complex, inhibiting TNF-α and IL-1β production in whole cells. Reduces death of crypt cells and improves gastrointestinal syndrome in mice.

Chemical Identifiers

CAS 1279713-77-7
Molecular Formula C18H13ClFNO3S
Molecular Weight (g/mol) 377.814
InChI Key IAASQMCXDRISAV-CYBMUJFWSA-N
Synonym cu cpt 4a, n-3-chloro-6-fluorobenzo b thien-2-yl carbonyl-d-phenylalanine, n-3-chloro-6-fluorobenzo b thiophene-2-yl carbonyl-d-phenylalanine, r-2-3-chloro-6-fluorobenzo b thiophene-2-carboxamido-3-phenylpropanoic acid
PubChem CID 53242268
IUPAC Name (2R)-2-[(3-chloro-6-fluoro-1-benzothiophene-2-carbonyl)amino]-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC(=O)C2=C(C3=C(S2)C=C(C=C3)F)Cl

Specifications

Quantity 50 mg
Formula Weight 377.82
Chemical Name or Material CU CPT 4a
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.