Learn More
Epredia™ CryoStar™ NX50 and NX70 Accessories
Enhance operation of Epredia™ CryoStar™ NX50 and NX70 Cryostats with Epredia™ CryoStar™ NX50 and NX70 Accessories.
16.40€ - 3310.00€
Products 12
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
12912735
View Documents
Epredia™
202180
Brush for cleaning vacuum hose
16.40€
Each
12902755
View Documents
Epredia™
334171
Brush, small beveled
61.60€
Each
12942755
View Documents
Epredia™
336080
Nozzle small (for sectioning)
209.00€
Each
12962765
View Documents
Epredia™
410540
Brush Shelf
221.00€
Each
12972765
View Documents
Epredia™
419500
Blade Handling Tool
188.00€
Each
12667576
View Documents
Epredia™
705840
Low Profile Blade Holder
3310.00€
Each
12902815
View Documents
Epredia™
705940
High Profile Blade Holder
3250.00€
Each
12932735
View Documents
Epredia™
281000
Coarse Filter
851.00€
Pack of 25
12912815
View Documents
Epredia™
705950
Steel Knife Carrier
2320.00€
Each
12912755
View Documents
Epredia™
334172
Magnetic Brush, Big This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
12932755
View Documents
Epredia™
334240
Funnel
56.40€
Each
12083049
View Documents
Epredia™
449980
Anti-Roll Plate Glass 69.5 mm
180.00€
Each
Description
Description
Enhance operation of Epredia™ CryoStar™ NX50 and NX70 Cryostats with Epredia™ CryoStar™ NX50 and NX70 Accessories.
Accessories Include:
  • Low Profile Blade Holder
  • High Profile Blade Holder
  • Steel Knife Holder
Documents
Product Certifications
Product Content Correction
Product Title
Epredia™ CryoStar™ NX50 and NX70 Accessories