Learn More
Spex CertiPrep™ Cryo-Stations
Product Code. 12567326 Shop All SPEX Certiprep Products
Click to view available options
For Use With (Equipment):
Cryo-Blocks
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 12567326
Brand: Spex CertiPrep™ 2600
This item requires a price update prior to being ordered.
Request a Price for this item here.
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Keep samples cold during sample manipulation and prior to or after grinding
A dedicated holding station for temperature-sensitive samples that must be kept chilled during processing with SPEX SamplePrep homogenisers. Closed system is filled with liquid nitrogen automatically or manually. Either sample vials and cryoblocks can be placed in the Cryostation to keep samples cold.
  • Holds two titre plates or three cryoblocks
  • Keep samples cold during sample manipulation and prior to or after grinding
Specifications
Product Type Cryo-Station
For Use With (Equipment) Cryo-Blocks
Includes Cutting board for preparing temperature sensitive samples
Model 2600