Learn More
Fisherbrand™ PTFE Magnetic Stir Bar, Cross-shaped
Use with containers, vessel, stirrers, and stirring hot plates. The PTFE-coated magnet stirrers are stable for many general purpose applications. The bars are cross-shaped for improved stirring action.
Brand:  Fisherbrand™ 001.2401
 View more versions of this product
Product Code. 11898932
  • 15.40€ / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Use with containers, vessel, stirrers, and stirring hot plates. The PTFE-coated magnet stirrers are stable for many general purpose applications. The bars are cross-shaped for improved stirring action.
  • Stable, general-purpose magnetic stirrer
  • Provides a deep, quiet vortex even at relatively high speeds
  • Autoclavable
Specifications
Specifications
Autoclavable
White
10 x 10 x 5mm
5mm
0.375 in.
PTFE
5/Pk.
PTFE Stirrer Bars Cross
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
Containers & Vessels / Stirrer Hot Plates
0.375 in.
10mm
10 mm
Cross