missing translation for 'onlineSavingsMsg'
Learn More

CP 93129 dihydrochloride, Tocris Bioscience™

Product Code. 18064042
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT1B agonist

Potent and highly selective 5-HT1B agonist (Ki values are 8.1, 1100, 1500, 2900 and 7200 nM for 5-HT1B, 5-HT1D, 5-HT1A, 5-HT1c and 5-HT2, respectively). Reduces food intake and decreases body weight in rats.

Chemical Identifiers

CAS 879089-64-2
Molecular Formula C12H17Cl2N3O2
Molecular Weight (g/mol) 306.187
InChI Key UXDAADMPPAHNMY-UHFFFAOYSA-N
Synonym cp-93129 dihydrochloride hydrate, 1,4-dihydro-3-1,2,3,6-tetrahydro-4-pyridinyl-5h-pyrrol 3,2-b pyridin-5-one dihydrochloride hydrate, cp-93129 dihydrochloride hydrate hplc, 3-1,2,3,6-tetrahydropyridin-4-yl-1h,4h-pyrrolo 3,2-b pyridin-5-one hydrate dihydrochloride
PubChem CID 71311960
IUPAC Name 3-(1,2,3,6-tetrahydropyridin-4-yl)-1,4-dihydropyrrolo[3,2-b]pyridin-5-one;hydrate;dihydrochloride
SMILES C1CNCC=C1C2=CNC3=C2NC(=O)C=C3.O.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 288.18
Percent Purity >98%
Chemical Name or Material CP 93129 dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'