missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP 154526, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, non-peptide CRF1 receptor antagonist (Ki values are 2.7 and > 10000 nM for CRF1 and CRF2 receptors respectively). Blocks CRF-induced adenylate cyclase activation (Ki = 3.7 nM) and attenuates activation of the HPA axis by CRF. Displays anxiolytic-like activity in the rat elevated plus-maze test; brain penetrant.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 400.99 |
Percent Purity | >99% |
Chemical Name or Material | CP 154526 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'