missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Cover Slips, Thickness 1.5, 18 x 24mm
Protect assay preparations from chemical attack using Epredia™ Cover Slips, Thickness 1.5, made of D 263 M colorless borosilicate glass.
Brand: Epredia™ BB01800240AC23MNZ0
30.45 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
Thickness 1.5:
- 0.16 to 0.19mm
Excellent Glass Quality:
- Virtually colorless appearance
- Very low levels of inclusions, striae, bubbles, streaks, etc.
- High spectral transmission
- Excellent flatness
- Finely adapted refractive index suited for microscopes
Ordering Information:
- Packed 100 pieces in plastic boxes
- Packing unit 1000 pieces
- Other sizes and thicknesses available on request
Specifications
24 mm | |
18 mm | |
#1.5 (0.16 to 0.19 mm) | |
18 x 24 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction