missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coronavirus Nucleoprotein Monoclonal Antibody (3878)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547531

Product Code. 17866447

  • 491.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specificity: Reactive with the NP of native SARS, MERS by IFA & COVID-19. Non-reactive with the following Coronavirus strains: 229E, NL63 & OC43. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

The coronavirus nucleocapsid (N) is a structural protein that forms complexes with genomic RNA, interacts with the viral membrane protein during virion assembly and plays a critical role in enhancing the efficiency of virus transcription and assembly. Amino acid sequence comparisons have shown that coronavirus nucleocapsid proteins have three distinct and highly conserved domains: two structural and independently folded structural regions, namely the N terminal domain (NTD/domain 1) and C-terminal domain (CTD/domain 3), which are separated by a intrinsically disordered central region (RNA-binding domain/domain 2); all three domains have been shown in different CoVs to bind with viral RNA. [PMID: 25105276].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Coronavirus Nucleoprotein
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Virus
Antibody
IgG1
ELISA, Immunocytochemistry
3878
Unconjugated
NP; SARS
Proprietary sequence.
100 μg
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.