Learn More
Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Translucent polymethylpentene; with polypropylene screw cap
Brand:  Corning™ 4990P-50
 View more versions of this product
Product Code. 17223554
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
Polypropylene
Narrow Mouth
1/Pk.
50 mL
10 mL
6/Cs.
Screw Cap