Learn More
Corning™ LSE™ Digital Dry Bath Accessory, Single Blocks
Shop All Corning Products
Click to view available options
For Use With (Equipment):
LSE Dry Bath Heater
Unit Size:
Each
Description
The single and dual block configurations are used in the Corning LSE Digital Dry Bath Heaters.
Specifications
Specifications
| Capacity (Metric) | 0.2 mL |
| For Use With (Equipment) | LSE Dry Bath Heater |
| Material | Stainless Steel |
| No. of Places | 48 |
| Type | Block |
| Tube Size | 0.2 mL |