Learn More
Corning™ Lambda Plus 8 and 12-Channel Pipettors
Alle ansehen Corning Produkte
Beschreibung
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
- Factory calibrated to strict ISO 8655 standards-3-year warranty
- Fully autoclavable and UV resistant
- Designed for universal fit with all common brands of pipet tips
- Adjustable tip ejector
Bestellinformation
Accessory stand and rack available separately (Cat. Nos. 07-201-790 and -789).
Serviceleistungen
3-Year
Spezifikation
Spezifikation
| Accuracy | ±3 to ±1% |
| Certifications/Compliance | CE |
| Autoclavable | Autoclavable |
| For Use With | Universal Pipette Tip |
| Number of Channels | 12 |
| Product Line | Lambda Plus |
| Sterility | Non-sterile |
| Type | Pipette |
| Volume (Metric) | 20 to 200 μL |
| UV Monitoring | Yes |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.