Learn More
Corning™ BioCoat™ Tumor Invasion Multiwell Plate
Product Code. 10007891
Click to view available options
No. of Wells:
24
96
Culture Area:
0.08 cm2
0.3 cm2
Quantity:
1
5
Unit Size:
Each
Pack of 5
Product Code. 10007891
Brand: Corning™ 354166
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running cell migration and invasion studies.
TRUSTED_SUSTAINABILITY
Specifications
Culture Area 0.3 cm2
Material PET
Pore Size 8 μm
Type Multiwell Plate
Quantity 5
No. per Pack 5/Pk.
No. per Case 5/Cs.
Surface Coating Matrigel™ Matrix
No. of Wells 24