Learn More
Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis
Product Code. 10778124
Click to view available options
No. of Wells:
24
96
Culture Area:
0.08 cm2
0.3 cm2
Quantity:
1
5
Unit Size:
Each
Pack of 5
Product Code. 10778124
Brand: Corning™ 354147
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes
A Falcon 24- and 96-Multiwell™ insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid
Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
TRUSTED_SUSTAINABILITY
Specifications
Culture Area 0.08 cm2
Pore Size 3 μm
Type Multiwell Insert Plate
For Use With (Application) High Throughput Screening
Quantity 1
Surface Coating Fibronectin
No. of Wells 96