Learn More
Serva Electrophoresis™ Coomassie™ Brilliant Blue G 250
43.56€ - 107.69€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11812020
|
Serva Electrophoresis™
1752401 |
25 g |
43.56€
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
11822020
|
Serva Electrophoresis™
1752402 |
100 g |
107.69€
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
Specifications
Dark Blue | |
Protein Stain |