Learn More
Gilac™ Cook Chill Crate
Cook Chill Crate is ideal for commercial kitchen use and the cold chain process, and is stacking and nesting when half turned.
Brand:  Gilac™ G180009-1
 View more versions of this product
Product Code. 17968581
  • 31.90€ / Each
Explore more special offers
Description
Description
  • 600 x 400 perforated crate.
  • Ideal for commercial kitchen use and the cold chain process.
  • Provides ventilation.
  • Stacking and nesting when half turned.
  • Large perforations meaning that temperatures can be brought down quickly and is easy to clean.
Specifications
Specifications
9.8 gal.
None
HDPE
Crate
600 mm
400 mm
197 mm
37 L
Red
Rectangular
600 x 400 x 197 mm
23.6 in.
15.7 in.
7.8 in.