Learn More
Gilac™ Cook Chill Crate
Cook Chill Crate is ideal for commercial kitchen use and the cold chain process, and is stacking and nesting when half turned.
Brand: Gilac™ G180121-1
This item is not returnable.
View return policy
Description
- 600 x 400 perforated crate.
- Ideal for commercial kitchen use and the cold chain process.
- Provides ventilation.
- Stacking and nesting when half turned.
- Large perforations meaning that temperatures can be brought down quickly and is easy to clean.
Specifications
7.1 gal. | |
None | |
HDPE | |
Crate | |
600 mm | |
400 mm | |
146 mm |
27 L | |
White | |
Rectangular | |
600 x 400 x 146 mm | |
23.6 in. | |
15.7 in. | |
5.7 in. |