Learn More
WTW™ Connection Cable for Depth Probes
For use with WTW™ MPP IDS depth probes with waterproof plug head.
Brand:  WTW™ 903852
 View more versions of this product
Product Code. 13466768
  • 164.00€ / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Also useful for plug head sensors
  • Available in a range of lengths
  • Supplied individually
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Connection Cable
6 m
MPP IDS with waterproof head sensors