missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard Solutions, Hanna Instruments™
A premium quality conductivity standard that is prepared against a NIST traceable potassium chloride solution.
Brand: Hanna Instruments HI70031P
27.87 EUR valid until 2025-04-18
Use promo code "24095" to get your promotional price.
This item is not returnable.
View return policy
Description
- Premium quality 1,413 μS/cm conductivity solution that is NIST traceable and contained in portable single-use sachets
- Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
- Convenient packaging for one time use
- Standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
- Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
- A choice of ready mixed calibration standards traceable to NIST
- Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air
Specifications
20 mL | |
Laboratory and Industrial Use | |
∼7 | |
Soluble in water |
1413 μS/cm | |
Colorless | |
∼100°C | |
Calibration Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Conductivity Standard Solutions, Hanna Instruments™ > mV Range: 1413uS/cm; Capacity: 20mL
Spot an opportunity for improvement?Share a Content Correction