Learn More
WTW™ Conductivity kit
For conductivity measuring systems, there are different check methods:The system including measuring cell can be calibrated with suitable traceably certifed standard solutions. An adjustment is usually not necessary due to the long-term stability of the used electrode materials. Special kits contain all necessary parts to test conductivity measuring systems according to pharamcopeia guidelines.
Brand: WTW™ 300569
Specifications
±2% | |
6R/SET/Lab1, LR 325/01 ultrapure water cond cell, D 01/T glass flow-through vessel | |
Conductivity meter series |
Conductivity kit | |
5μS/cm |