Learn More
Olympus™ Condenser, Long Working Distance, U-LWCO
Long working distance condenser. Olympus™ Condenser, Long Working Distance, U-LWCO works with BX, IX, and MX microscopes.
Brand:  Olympus™ K10017635
Product Code. 12686692
  • 1215.00€ / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Long working distance: 12mm.
Specifications
Specifications
BX, IX, and MX microscopes
0.65