missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Cond 3210™ Portable Conductivity Meter
Brand: WTW™ 2CA201
This item is not returnable.
View return policy
Description
Specifications
Meter with TetraCon 325 conductivity cell | |
±0.5% Full-scale ±1 LSD | |
Fixed: 0.475cm–1/1cm–1, Calibratable: 0.450 to 0.500cm–1/0.585 to 715cm–1/0.800 to 1.200cm–1 | |
0 μS to 1000 mS | |
Automatic | |
0.1 ppt | |
0 to 200 ppt | |
±0.5% Full-scale | |
±0.1°C | |
200 | |
IP67 | |
3 Years | |
4 x 1.5V AA or 4 x 1.2V NiMH | |
Portable Conductivity Meter | |
7.9 cm | |
NiMH |
Meter with carry case including 4-electrode conductivity cell TetraCon™ 325, standard solution 1413μS/cm at 25°C, stand, beaker, instruction manual, CD-ROM and batteries. | |
For professional field measurements requiring documentation and data transfer | |
1, 2, 3, 4, 5-point WTW Technical NIST and 20 additional Buffer sets | |
0.01, 0.1 mS | |
0 to 70 ppt | |
±0.2% Full-scale | |
0.1 ppt | |
0.1 | |
LCD | |
CE | |
0.6 kg | |
Cond 3210 | |
-5°C to +105°C | |
17.7 cm | |
5.7 cm | |
Up to 1000 hr. |