missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Cond 3210™ Portable Conductivity Meter

Brand:  WTW™ 2CA204

Product Code. 10095283

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Meter with LR325/01 conductivity cell
±0.5% Full-scale ±1 LSD
Fixed: 0.475cm–1/1cm–1, Calibratable: 0.450 to 0.500cm–1/0.585 to 715cm–1/0.800 to 1.200cm–1
0 μS to 1000 mS
Automatic
0.1 ppt
0 to 200 ppt
±0.5% Full-scale
±0.1°C
200
IP67
3 Years
4 x 1.5V AA or 4 x 1.2V NiMH
Portable Conductivity Meter
7.9 cm
NiMH
Meter with carry case including 2-electrode conductivity cell LR 325/01 including flow-through vessel, stand, beaker, short instruction manual, CD-ROM and batteries
For professional field measurements requiring documentation and data transfer
1, 2, 3, 4, 5-point WTW Technical NIST and 20 additional Buffer sets
0.01, 0.1 mS
0 to 70 ppt
±0.2% Full-scale
0.1 ppt
0.1
LCD
CE
0.6 kg
Cond 3210
-5°C to +105°C
17.7 cm
5.7 cm
Up to 1000 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions