Learn More
Cole-Parmer™ Compression to Male Threaded Adapter
Compression fittings are an excellent choice for connecting rigid tubing systems
Brand: Cole-Parmer™ 06385-87
This item is not returnable.
View return policy
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- PVDF Composition
Specifications
Compression to Male Threaded Adapter | |
Autoclavable | |
1 5/8 in. | |
1/2 in. | |
1/2 in. NPT(M) | |
24 mm | |
Straight |
Compression x NPT(M) Connection | |
PVDF | |
10 Fittings | |
1/2 in. | |
Autoclave or Ethylene Oxide | |
15/16 in. |