Learn More
Nabertherm™ Compact Tube Furnace with P580 Controller
Product Code. p-7236235 Shop All Nabertherm Products
Click to view available options
Diameter (Metric) Process Tube:
50 mm
120 mm
170 mm
Heated Zone:
500 mm
750 mm
1000 mm
Temperature (Metric):
1200°C
1300°C
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17398079
Brand: Nabertherm™ R252W2AN3
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Controller P580 with 50 freely adjustable programs and 40 segments for three-zone operation
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Specifications
Controller Type P580
Heated Zone 500 mm
Temperature (Metric) 1200°C
Type Compact Tube Furnace
Dimensions (L x W x H) Exterior 670 x 340 x 508 mm
Length (Metric) 700 mm
Diameter (Metric) Process Tube 50 mm