Learn More
WTW™ Combined Fluoride Selective Electrode
The combined electrodes type 800 are excellent in their handling and offer the additional opportunity to also measure small sample volumes, as the ion selective and the reference electrodes are combined in a compact rod. Furthermore, they are convincing with their exceptionally great price/performance ratio.
Brand:  WTW™ 106667
Product Code. 10163482
  • 1355.00€ / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Measuring small sample volumes
DIN plug
Combined Fluoride Selective Electrode