missing translation for 'onlineSavingsMsg'
Learn More

CombiCheck™ 70 Standard Solution Ammonium, COD, Nitrogen, WTW™

Product Code. 10355611
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Physical Form Liquid
Concentration Ammonium 50mg/L , COD 5000mg/L ,Nitrogen 50mg/L
Solution Contains Ammonium, COD, Nitrogen
Color Colorless
Quantity 100 mL
Solubility Soluble in water
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'