Learn More
BUCHI Combi-Clip
For use with Rotavapor R-300
Brand:  BUCHI 11059770
Product Code. 15459092
  • 51.00€ / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Patented Combi-Clip with snap lock mechanism
  • For fastening evaporating flask on to vapor duct
  • Vapor duct sold separately
Specifications
Specifications
PBT
Combi Clip
Fastening Evaporating Flask on Vapor Duct
Rotavapor R-300
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BUCHI Combi-Clip > Combi-Clip