missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ AF325 Platinum-Cobalt/Hazen/APHA Test Kit

The Lovibond™ AF325 is a test kit for visual colour grading of Platinum-Cobalt/Hazen/APHA Colour over the range 10 to 250mg Pt/l.

Brand:  Lovibond™ 433250

Additional Details : Weight : 5.00000kg

Product Code. 10732551

  • 1840.00€ / Each
Estimated Shipment: 30-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Récupel-bijdrage uitgesloten

Description

Description

  • Typically used for lightly coloured liquids such as ground water and chemicals
  • Colour is determined by visually matching a column of sample in a Nessler cylinder with precalibrated coloured glass standards, which are stable-coloured and not affected by UV light or extreme environmental conditions
  • Because sample colours are relatively unsaturated, the kit employs a long path length Nessleriser comparator to give greater measurement sensitivity and accuracy
Specifications

Specifications

ASTM D1209
Applications: Beers, Malts and Caramels, Food and Beverages, Household Products, Medical and Clinical, Petroleum Oils and Waxes, Edible Oils and Fats, Sugar Solutions, Syrups and Honey, Pharmaceuticals and Cosmetics, Textiles, Coatings, Plastics, Chemicals, Swimming Pools and Spas, Water and Waste Waters
AF325
Color Measurement Test Kit
113mm Nessleriser 2150, daylight 2000 lighting unit, stand, two AF306/P, Pt-Co/Hazen disc NSH, Pt-Co/Hazen disc NSB and Nessleriser stand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions