missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coenzyme A Trilithium Salt Dihydrate ≥96%, White Crystalline Powder, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
250 mg
500 mg
Unit Size:
100mg
10mg
250mg
25mg
500mg
50mg
Specifications
Specifications
| Color | White |
| Assay Percent Range | ≥96% |
| Quantity | 25 mg |
| Formula Weight | 821.4 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Coenzyme A Trilithium Salt Dihydrate >96% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction