missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ COD Test Tubes

182.00€ - 190.00€

Specifications

Description COD Test Tube
Volume (Metric) 0 to 15,000 mg/L
For Use With (Application) For Wastewater Treatment Plants and Food Manufacturing Facilities
Quantity 25 Pack
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
11758763
View Documents
Hanna Instruments™
HI93754B25
Accuracy: 4%
182.00€
87.50mL
Please to purchase this item. Need a web account? Register with us today!
11768763
View Documents
Hanna Instruments™
HI93754C25
Accuracy: 3%
190.00€
125mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

COD Test Tube
0 to 15,000 mg/L
For Wastewater Treatment Plants and Food Manufacturing Facilities
25 Pack
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • COD MR reagent

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P391-Collect spillage.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, mercury (II) sulphate, potassium dichromate

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.