Learn More
Aalborg™ CO2 Direct Flow Tube
Aalborg™ CO2 Direct Flow Tube completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
73.00€ - 118.00€
Specifications
Type CO2 Direct Flow Tube
View More Specs
Products 4
Product Code Brand Airflow Rate Width (Metric) Price Quantity & Availability  
Product Code Brand Airflow Rate Width (Metric) Price Quantity & Availability  
11882270
View accessory products View Documents
Aalborg™
FT-014-18-GL-VN
6 L/min. 65 mm
73.00€
Each
Estimated Shipment: 26-02-2025
to see stock.
Add to basket
11862250
View accessory products View Documents
Aalborg™
FT-052-14-GL-VN
1 L/min. 65 mm
73.00€
Each
Estimated Shipment: 26-02-2025
to see stock.
Add to basket
11842450
View accessory products View Documents
Aalborg™
FT-062-09-GL-VN
100 mL/min. 150 mm
118.00€
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
11882450
View accessory products View Documents
Aalborg™
FT-062-14-ST-VN
300 mL/min. 150 mm
118.00€
Each
Estimated Shipment: 26-02-2025
to see stock.
Add to basket
Description
Description
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Specifications
CO2 Direct Flow Tube