missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ CN 60 Ionselective Combination Electrode for Cyanide
CN 60 Ionselective combination solid state body electrode is for determination of Cyanide-ions.
Brand: SI Analytics™ 285130390
This item is not returnable.
View return policy
Description
- CN 60
- For the determination of Cyanide-ions, solid state body electrode
- Temp.range: 0°C to 80°C
- pH-range: 0 to 14
- Measuring range: 0.2 to 260 mg/L
- IONSELEKT. COMB. ELECTRODE CN 60
Specifications
Cyanide | |
Solid State | |
Electrode |
0 to 14 | |
0°C to 80°C | |
0.2 to 260 mg/L |