Learn More
1H-Benzotriazole-(ring-d4) solution, Honeywell Fluka™
10 μg/mL in acetone, analytical standard, reference material
Brand:  Honeywell Fluka R1165-5ML
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15901101
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1185072-03-0, 67-64-1
5 mL
0.79 g/cm3
-17°C
Liquid
Glass Bottle
Analytical Standard
55°C to 57°C
Parabenes