missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clenbuterol-d9, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
| Packaging | Glass Bottle |
| CAS | 129138-58-5 |
| Physical Form | Solid |
| Industry Type | Beta-agonists and Antagonists |
| Quantity | 10 mg |
| Grade | Analytical Standard |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido